missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC28A2 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
201.00 € - 481.00 €
Specifications
| Antigen | SLC28A2 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18623431
|
Novus Biologicals
NBP2-93794-0.02ml |
0.02 mL |
201.00 €
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18654991
|
Novus Biologicals
NBP2-93794-0.1ml |
0.1 mL |
481.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SLC28A2 Polyclonal antibody specifically detects SLC28A2 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| SLC28A2 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Endocrinology, Signal Transduction | |
| PBS (pH 7.3), 50% glycerol | |
| 9153 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| CNT2SPNT, Concentrative nucleoside transporter 2, FLJ21468, HCNT2, HsT17153, MGC138252, Na(+)/nucleoside cotransporter 2, sodium/nucleoside cotransporter 2, Sodium/purine nucleoside co-transporter, Sodium-coupled nucleoside transporter 2, solute carrier family 28 (sodium-coupled nucleoside transporter), member 2, Solute carrier family 28 member 2, SPNT1CNT 2 | |
| A synthetic peptide corresponding to a sequence within amino acids 350-450 of human SLC28A2 (NP_004203.2). GAFIAFGVDASSLISASVMAAPCALASSKLAYPEVEESKFKSEEGVKLPRGKERNVLEAASNGAVDAIGLATNVAANLIAFLAVLAFINAALSWLGELVDI | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title