missing translation for 'onlineSavingsMsg'
Learn More

SLC43A3 Antibody, Novus Biologicals™

Product Code. p-200060440 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
25 μL
Unit Size:
100µL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18686708 25 μL 25µL
18698747 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18686708 Supplier Novus Biologicals Supplier No. NBP26903225ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SLC43A3 Polyclonal antibody specifically detects SLC43A3 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC43A3
Applications Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL
Formulation PBS (pH 7.2) and 40% Glycerol
Gene Alias DKFZp762A227, EEG1, FOAP-13, likely ortholog of mouse embryonic epithelial gene 1, PRO1659, Protein FOAP-13, SEEEG-1, solute carrier family 43 member 3, solute carrier family 43, member 3
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to amino acids: FKNEDYFKDLCGPDAGPIGNATGQADCKAQDERFS
Purification Method Protein A purified
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29015
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.