missing translation for 'onlineSavingsMsg'
Learn More

SLC8A2 Antibody, Novus Biologicals™

Product Code. p-200057957 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
25 μL
Unit Size:
0.1mL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18646526 25 μL 25µL
18689628 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18646526 Supplier Novus Biologicals Supplier No. NBP24914525ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SLC8A2 Polyclonal antibody specifically detects SLC8A2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC8A2
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500
Formulation PBS (pH 7.2), 40% Glycerol
Gene Alias KIAA1087, Na(+)/Ca(2+)-exchange protein 2, Na+/Ca2+-exchanging protein Nac2, NCX2, sodium/calcium exchanger 2, sodium-calcium exchanger 2, solute carrier family 8 (sodium/calcium exchanger), member 2, solute carrier family 8 (sodium-calcium exchanger), member 2, solute carrier family 8 member 2
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to amino acids: AGEDEDDGASRIFFEPSLYHCLENCGSV
Purification Method Immunogen affinity purified
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6543
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.