missing translation for 'onlineSavingsMsg'
Learn More

Slug Antibody, Novus Biologicals™

Product Code. 18748973 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
Unit Size:
0.1mg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18748973 0.1 mg 0.1mg
18771734 0.025 mg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18748973 Supplier Novus Biologicals Supplier No. NBP227182

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody has been used in 3 publications

Slug Polyclonal specifically detects Slug in Human, Mouse, Canine, Equine samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Dilution Western Blot 1:2000, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200
Host Species Rabbit
Immunogen Amino acids 82-110 of human SNAIL2 (SNAI2) were used as the immunogen.
Molecular Weight of Antigen 30 kDa
Quantity 0.1 mg
Regulatory Status RUO
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.