missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Smad4 Monoclonal antibody specifically detects Smad4 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | Smad4 |
| Applications | Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | CL9774 |
| Conjugate | DyLight 755 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | DPC4SMAD, mothers against DPP homolog 4 (Drosophila), hSMAD4, JIP, MAD homolog 4, MADH4mothers against decapentaplegic homolog 4, mothers against decapentaplegic homolog 4, mothers against decapentaplegic, Drosophila, homolog of, 4, SMAD 4, SMAD family member 4MAD, mothers against decapentaplegic homolog 4 (Drosophila), SMAD, mothers against DPP homolog 4, Smad4 |
| Host Species | Mouse |
| Immunogen | This antibody was developed using a recombinant protein derived from Q13485, with the exact immunogen sequence remaining proprietary. |
| Purification Method | Protein A purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?