missing translation for 'onlineSavingsMsg'
Learn More

SMARCA1 Antibody [DyLight 550], Novus Biologicals Biologicals™

Product Code. 30501906 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30501906 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30501906 Supplier Novus Biologicals Supplier No. NBP335110R

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SMARCA1 Polyclonal antibody specifically detects SMARCA1 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Immunoprecipitation,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen SMARCA1
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence
Classification Polyclonal
Conjugate DyLight 550
Formulation 50mM Sodium Borate
Gene Alias ATP-dependent helicase SMARCA1, DKFZp686D1623, EC 3.6.1, EC 3.6.4.-, FLJ41547, global transcription activator homologous sequence, Nucleosome-remodeling factor subunit SNF2L, NURF140, probable global transcription activator SNF2L1, SNF2L, SNF2L1ISWI, SNF2LB, SNF2-like 1, SNF2LT, subfamily a, member 1, sucrose nonfermenting 2-like protein 1, SWI, SWI/SNF related, matrix associated, actin dependent regulator of chromatin, SWI/SNF-related matrix-associated actin-dependent regulator of chromatin a1, SWI/SNF-related matrix-associated actin-dependent regulator of chromatinsubfamily A member 1, SWI2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-200 of human SMARCA1 (NP_003060.2).,, Sequence:, MEQDTAAVAATVAAADATATIVVIEDEQPGPSTSQEEGAAAAATEATAATEKGEKKKEKNVSSFQLKLAAKAPKSEKEMDPEYEEKMKADRAKRFEFLLKQTELFAHFIQPSAQKSPTSPLNMKLGRPRIKKDEKQSLISAGDYRHRRTEQEEDEELLSESRKTSNVCIRFEVSPSYVKGGPLRDYQIRGLNWLISLYEN
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6594
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.