missing translation for 'onlineSavingsMsg'
Learn More

SMC3 Antibody, DyLight 350, R&D Systems™

Product Code. 18608199 Shop All Bio Techne Products
Change view
Click to view available options
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Unit Size
18608199 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18608199 Supplier Novus Biologicals Supplier No. NB100207UV
  • ONLINE EXCLUSIVE PRICE
    720.26 €
    632.10 € / 0.1mL
    Save 88.16 €
    12% Off
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Rabbit Polyclonal Antibody

SMC3 Polyclonal specifically detects SMC3 in C. elegans, Human, Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SMC3
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation
Formulation 50mM Sodium Borate
Gene Alias Bamacan, BAMSMC protein 3, Basement membrane-associated chondroitin proteoglycan, BMH, Chondroitin sulfate proteoglycan 6, chondroitin sulfate proteoglycan 6 (bamacan), Chromosome-associated polypeptide, CSPG6structural maintenance of chromosomes protein 3, hCAP, HCAPbamacan, SMC-3, SMC3L1CDLS3, structural maintenance of chromosomes 3
Target Species C. elegans, Human, Mouse
Product Type Antibody
Product Line Novus
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.