missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SMN Antibody (2B1), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 21 publication
Brand: Novus Biologicals NB100-1936
This item is not returnable.
View return policy
Description
SMN Monoclonal specifically detects SMN in Human, Mouse, Rat, Primate, Xenopus, Zebrafish samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications
| SMN | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 2 ug/ml, Flow Cytometry reported in scientific literature (PMID 30102724), ELISA reported in scientific literature (PMID 23973875), Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:250, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:200 | |
| Gemin-1, Kugelberg-Welander disease), SMNC, survival motor neuron protein, survival of motor neuron 1, telomeric | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, mouse, rat, monkey and Xenopus. Other species not tested. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| 2B1 | |
| Unconjugated | |
| Q16637, P97801, O35876 | |
| SMN1 | |
| Purified recombinant His6-tagged human SMN protein. [Swiss-Prot# Q16637] | |
| 0.1 mL | |
| Apoptosis, Neurodegeneration, Neuroscience, Transcription Factors and Regulators | |
| 6606 | |
| Human, Mouse, Rat, Primate, Xenopus species | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction