missing translation for 'onlineSavingsMsg'
Learn More

SMN Antibody (2B1), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 21 publication

190.00 € - 594.00 €

Specifications

Antigen SMN
Clone 2B1
Concentration 1.0 mg/mL
Dilution Western Blot 2 ug/ml, Flow Cytometry reported in scientific literature (PMID 30102724), ELISA reported in scientific literature (PMID 23973875), Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:250, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:200
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18064984
View Documents
Novus Biologicals
NB100-1936SS
0.025 mL
190.00 €
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18788073
View Documents
Novus Biologicals
NB100-1936
0.1 mL
594.00 €
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

SMN Monoclonal specifically detects SMN in Human, Mouse, Rat, Primate, Xenopus, Zebrafish samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications

Specifications

SMN
1.0 mg/mL
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
Unconjugated
Mouse
Apoptosis, Neurodegeneration, Neuroscience, Transcription Factors and Regulators
Q16637, P97801, O35876
6606
Purified recombinant His6-tagged human SMN protein. [Swiss-Prot# Q16637]
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
2B1
Western Blot 2 ug/ml, Flow Cytometry reported in scientific literature (PMID 30102724), ELISA reported in scientific literature (PMID 23973875), Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:250, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:200
Monoclonal
Purified
RUO
Human, Mouse, Rat, Primate, Xenopus species
Gemin-1, Kugelberg-Welander disease), SMNC, survival motor neuron protein, survival of motor neuron 1, telomeric
SMN1
IgG1
Protein G purified
Human, mouse, rat, monkey and Xenopus. Other species not tested.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.