missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ SNAP25 Antibody (B-29-F9), Novus Biologicals™

Product Code. 30066935 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30066935 25 μg 25µg
30066833 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30066935 Supplier Novus Biologicals™ Supplier No. NBP33085425ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

SNAP25 Monoclonal antibody specifically detects SNAP25 in Bovine,Chicken,Human,Monkey,Mouse,Rat samples. It is validated for Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen SNAP25
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone B-29-F9
Conjugate Unconjugated
Dilution Western Blot 1-5 μg/mL, Immunohistochemistry-Paraffin
Formulation Phosphate buffered saline
Gene Alias bA416N4.2, dJ1068F16.2, FLJ23079, resistance to inhibitors of cholinesterase 4 homolog, RIC4, RIC-4, SEC9, SNAP-25SNAP, SUP, Super protein, Synaptosomal-associated 25 kDa protein, synaptosomal-associated protein 25, synaptosomal-associated protein, 25kD, synaptosomal-associated protein, 25kDa
Host Species Mouse
Immunogen Hybridoma produced by the fusion of splenocytes from BALB/c mice immunized with purified rat SNAP25 and mouse NS1 cells.
Purification Method Protein A or G purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Membrane Trafficking and Chaperones, Neuronal Cell Markers, Neuroscience, Neurotransmission, Vision
Primary or Secondary Primary
Gene ID (Entrez) 6616
Target Species Bovine, Chicken, Human, Monkey, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.