Learn More
SNX15 Mouse, anti-Human, CoraLite Plus 488, Clone: 1A10B7, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-68239-100UL
Description
The SNX15 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human SNX15. This antibody recognizes human antigen. The SNX15 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
| SNX15 | |
| 1A10B7 | |
| CoraLite Plus 488 | |
| Mouse | |
| Primary | |
| Human | |
| 155 kDa | |
| IgG2a |
| Immunofluorescence | |
| 1000 μg/mL | |
| HSAF001435, SNX15, sorting nexin 15 | |
| 100 μL | |
| 29907 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.