missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SNX15 Mouse, anti-Human, CoraLite Plus 488, Clone: 1A10B7, Proteintech
Mouse Monoclonal Antibody
145.00 € - 450.00 €
Specifications
| Antigen | SNX15 |
|---|---|
| Clone | 1A10B7 |
| Concentration | 1000 μg/mL |
| Applications | Immunofluorescence |
| Conjugate | CoraLite Plus 488 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18018105
|
Proteintech
CL488-68239-20UL |
20 μL |
145.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18088115
|
Proteintech
CL488-68239-100UL |
100 μL |
450.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The SNX15 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human SNX15. This antibody recognizes human antigen. The SNX15 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
| SNX15 | |
| 1000 μg/mL | |
| CoraLite Plus 488 | |
| Mouse | |
| 29907 | |
| IgG2a | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. |
| 1A10B7 | |
| Immunofluorescence | |
| Liquid | |
| Human | |
| HSAF001435, SNX15, sorting nexin 15 | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title