missing translation for 'onlineSavingsMsg'
Learn More

SOCS-1 Antibody, Novus Biologicals™

Product Code. 18666925 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
200 μg
50 μg
Unit Size:
100µL
200µL
50µL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18666925 200 μg 200µL
18608325 100 μg 100µL
18687625 50 μg 50µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18666925 Supplier Novus Biologicals Supplier No. NBP298892200ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SOCS-1 Polyclonal antibody specifically detects SOCS-1 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen SOCS-1
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunocytochemistry/ Immunofluorescence 1:300-1:10000, Immunohistochemistry-Paraffin 1:500-1:2000
Formulation 0.2 um filtered solution in PBS
Gene Alias JABTec-interacting protein 3, JAK binding protein, JAK-binding protein, SOCS-1CIS1, SSI-1CISH1, SSI1cytokine-inducible SH2 protein 1, STAT induced SH3 protein 1, STAT-induced STAT inhibitor 1, suppressor of cytokine signaling 1, TIP-3, TIP3Cish1
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human SOCS-1.
Purification Method Antigen and protein A Affinity-purified
Quantity 200 μg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 8651
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.