missing translation for 'onlineSavingsMsg'
Learn More

SOD1/Cu-Zn SOD Antibody, Novus Biologicals™

Product Code. 18718353 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mg
Unit Size:
0.1mg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18718353 0.025 mg 25µg
18771274 - 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18718353 Supplier Novus Biologicals Supplier No. NBP224915SS

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody has been used in 9 publications

SOD1/Cu-Zn SOD Polyclonal specifically detects SOD1/Cu-Zn SOD in Human, Mouse, Rat, Bovine samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Classification Polyclonal
Host Species Rabbit
Quantity 0.025 mg
Regulatory Status RUO
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.