missing translation for 'onlineSavingsMsg'
Learn More

Sodium 3-Sulfobenzoate, Mikromol™

Product Code. 30049199
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30049199 100 mg 100mg
30050121 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30049199 Supplier Mikromol Supplier No. MM0484.01
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Sodium 3-Sulfobenzoate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sodium 3-Sulfobenzoate
CAS 17625-03-5
Analyte or Component Names Sodium 3-Sulfobenzoate
Shipping Condition Room Temperature
Molecular Formula C7 H5 O5 S . Na
Certifications/Compliance ISO 17025
Synonym Sodium 3-Sulfobenzoate
InChI Formula InChI=1S/C7H6O5S.Na/c8-7(9)5-2-1-3-6(4-5)13(10,11)12;/h1-4H,(H,8,9)(H,10,11,12);/q;+1/p-1
SMILES [Na+].OC(=O)c1cccc(c1)S(=O)(=O)[O-]
IUPAC Name sodium;3-carboxybenzenesulfonate
Molecular Weight (g/mol) 224.17
Quantity 100 mg
Formula Weight 223.9755 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.