missing translation for 'onlineSavingsMsg'
Learn More

Sodium Benzoate, Mikromol™

Product Code. 17921786
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17921786 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17921786 Supplier Mikromol Supplier No. MM0730.00
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Sodium Benzoate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Benzoic Acid Sodium Salt
CAS 532-32-1
Analyte or Component Names Benzoic Acid Sodium Salt
Shipping Condition Room Temperature
Molecular Formula C7 H5 O2 . Na
Certifications/Compliance ISO 17034
Synonym Sodium Benzoate, Benzoic acid sodium salt (1:1), Benzoic acid sodium salt, Sodium benzoate, Antimol, Benzoate of soda, E 211, E 221, E 221 (nucleating agent), Fuminaru, Purox S, Sobenate, SKF-30573Z
InChI Formula InChI=1S/C7H6O2.Na/c8-7(9)6-4-2-1-3-5-6;/h1-5H,(H,8,9);/q;+1/p-1
SMILES [Na+].[O-]C(=O)c1ccccc1
IUPAC Name sodium;benzoate
Molecular Weight (g/mol) 144.1
Quantity 500 mg
Formula Weight 144.0187 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.