missing translation for 'onlineSavingsMsg'
Learn More

Sodium Cromoglicate, Mikromol™

Product Code. 17944456
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17944456 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17944456 Supplier Mikromol Supplier No. MM0039.00
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Sodium Cromoglicate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sodium cromoglicate
CAS 15826-37-6
Analyte or Component Names Sodium cromoglicate
Shipping Condition Room Temperature
Molecular Formula C23 H14 O11 . 2 Na
Certifications/Compliance ISO 17034
Synonym Sodium Cromoglicate, 5,5'-[(2-Hydroxy-1,3-propanediyl)bis(oxy)]bis[4-oxo-4H-1-benzopyran-2-carboxylic acid sodium salt (1:2), 5,5'-[(2-Hydroxy-1,3-propanediyl)bis(oxy)]bis[4-oxo-4H-1-benzopyran-2-carboxylic acid disodium salt, 5,5'-[(2-Hydroxytrimethylene)dioxy]bis[4-oxo-4H-1-benzopyran-2-carboxylic acid disodium salt, 1,3-Bis(2-carboxychromon-7-yloxy)-2-hydroxypropane disodium salt, Aarane, Alercrom, Alerion, Allergocrom, Altoderm, Colimune, Crolom, Cromoglycate, Cromoglycate disodium, Cromolyn disodium salt, Cromolyn sodium, Cromovet, DNCG, DSCG, Disodium 1,3-bis(2-carboxy-5-chromonyloxy)-2-propanol, Disodium 1,3-bis(2-carboxychromon-5-yloxy)-2-hydroxypropane, Disodium 1,3-bis(2-carboxychromon-5-yloxy)propan-2-ol, Disodium 5,5'-[(2-hydroxytrimethylene)dioxy]bis[4-oxo-4H-1-benzopyran-2-carboxylate], Disodium chromoglycate, Disodium cromoglycate, FPL 670, Fisonair, Fivent, Gastrocrom, Gastrofrenal, Inostral, Intal, Introl, Irtan, Lomudal, Lomupren, Lomusol, Lomuspray, Nalcrom, Nalcron, NasalCrom, Nasmil, Opticrom, Rynacrom, Sodium cromoglycate, Sodium cromolyn, Sofro, Vividrin
InChI Formula InChI=1S/C23H16O11.2Na/c24-11(9-31-14-3-1-5-16-20(14)12(25)7-18(33-16)22(27)28)10-32-15-4-2-6-17-21(15)13(26)8-19(34-17)23(29)30;;/h1-8,11,24H,9-10H2,(H,27,28)(H,29,30);;/q;2*+1/p-2
SMILES [Na+].[Na+].OC(COc1cccc2OC(=CC(=O)c12)C(=O)[O-])COc3cccc4OC(=CC(=O)c34)C(=O)[O-]
IUPAC Name disodium;5-[3-(2-carboxylato-4-oxochromen-5-yl)oxy-2-hydroxypropoxy]-4-oxochromene-2-carboxylate
Molecular Weight (g/mol) 512.33
Quantity 500 mg
Formula Weight 512.0332 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.