missing translation for 'onlineSavingsMsg'
Learn More

Reduced Folate Carrier/SLC19A1 Antibody (890513) [FITC], R&D Systems™

Product Code. 18606574 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18606574 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18606574 Supplier Novus Biologicals Supplier No. FAB8450F
  • ONLINE EXCLUSIVE PRICE
    728.45 €
    627.90 € / 0.1mL
    Save 100.55 €
    13% Off
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Mouse Monoclonal Antibody

Reduced Folate Carrier/SLC19A1 Monoclonal specifically detects Reduced Folate Carrier/SLC19A1 in Human samples. It is validated for CyTOF, Flow Cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Reduced Folate Carrier/SLC19A1
Applications CyTOF, Flow Cytometry
Classification Monoclonal
Clone 890513
Conjugate FITC
Dilution Flow Cytometry, CyTOF-ready
Formulation PBS
Gene Alias CHMD, FLOT1, folate transporter 1, FOLThuman reduced folate carrier (RFC)10Intestinal folate carrier 1, IFC1, IFC-1, Placental folate transporter, Reduced folate carrier protein, REFC, RFC, RFC1, solute carrier family 19 (folate transporter), member 1, Solute carrier family 19 member 1
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human Reduced Folate Carrier/SLC19A1, Met1-Gln591, Accession # P41440
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6573
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG2a
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.