missing translation for 'onlineSavingsMsg'
Learn More

Sodium Methyl Parahydroxybenzoate, LoGiCal

Product Code. 30463575
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30463575 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30463575 Supplier LoGiCal Supplier No. LGCFOR0431.01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Specifikationer

Chemical Name or Material Sodium Methyl Parahydroxybenzoate
CAS 5026-62-0
Analyte or Component Names Sodium Methyl Parahydroxybenzoate
Shipping Condition Room Temperature
Molecular Formula C8 H7 O3 . Na
Certifications/Compliance ISO 9001
Synonym Sodium Methyl Parahydroxybenzoate, SKF-4382-Z
InChI Formula InChI=1S/C8H8O3.Na/c1-11-8(10)6-2-4-7(9)5-3-6;/h2-5,9H,1H3;/q;+1/p-1
Solution Type Neat
SMILES [Na+].COC(=O)c1ccc([O-])cc1
IUPAC Name sodium;4-methoxycarbonylphenolate
Industry Type Clinical Testing
Molecular Weight (g/mol) 174.13
Quantity 10 mg
Formula Weight 174.0293 g/mol
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.