missing translation for 'onlineSavingsMsg'
Learn More

Sodium phenoxide, 98%

Product Code. 11441597
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11441597 250 g 250g
11431597 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11441597 Supplier Thermo Scientific Alfa Aesar Supplier No. B25002.30
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 139-02-6 | C6H5NaO | 116.10 g/mol

Sodium phenoxide is used as a precursor to aryl ethers and involved in the preparation of phenyl ethers and metal phenolates. It serves as an antiseptic and used in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium phenoxide is used as a precursor to aryl ethers and involved in the preparation of phenyl ethers and metal phenolates. It serves as an antiseptic and used in organic synthesis.

Solubility
Very soluble in water, soluble in alcoholSoluble in water, acetone and alcohol.

Notes
Hygroscopic and air sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 139-02-6
Molecular Formula C6H5NaO
Molecular Weight (g/mol) 116.10
MDL Number MFCD00013134
InChI Key NESLWCLHZZISNB-UHFFFAOYSA-M
Synonym sodium phenoxide, sodium phenolate, sodium phenate, phenolate sodium, sodium benzenolate, sodium carbolate, phenol sodium, phenol sodium salt, phenol, sodium salt, naoph
PubChem CID 4445035
ChEBI CHEBI:52476
SMILES [Na+].[O-]C1=CC=CC=C1

Specifications

Melting Point >300°C
Odor Phenol-like
Linear Formula C6H5ONa
Quantity 250 g
UN Number UN2905
Sensitivity Air Sensitive
Solubility Information Very soluble in water,soluble in alcoholSoluble in water,acetone and alcohol.
IUPAC Name sodium benzenolate
Formula Weight 116.09
Percent Purity 98%
Chemical Name or Material Sodium phenoxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.