Learn More
Thermo Scientific Chemicals Sodium phosphate, 0.2M buffer soln., pH 7.0
86.40 € - 110.00 €
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15425709
|
Thermo Scientific Chemicals
J63482.AP |
500 mL |
86.40 €
500mL |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15435709
|
Thermo Scientific Chemicals
J63482.K2 |
1 L |
110.00 €
1 L |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Sodium phosphate is widely used as a reagent with high buffering capacity for molecular biology, biochemistry, and chromatography
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsSodium phosphate is widely used as a reagent with high buffering capacity for molecular biology, biochemistry, and chromatography
Solubility
Fully miscible.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only