missing translation for 'onlineSavingsMsg'
Learn More

Sodium Picosulfate, Mikromol™

Product Code. 17991466
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17991466 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17991466 Supplier Mikromol Supplier No. MM0054.000250
 Restricted Item
Estimated Shipment: 05-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Sodium Picosulfate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sodium Picosulfate
CAS 10040-45-6
Analyte or Component Names Sodium Picosulfate
Shipping Condition Room Temperature
Molecular Formula C18 H13 N O8 S2 . 2 Na
Certifications/Compliance ISO 17034
Synonym Sodium Picosulfate, Phenol, 4,4'-(2-pyridinylmethylene)bis-, 1,1'-bis(hydrogen sulfate), sodium salt (1:2), Phenol, 4,4'-(2-pyridinylmethylene)bis-, bis(hydrogen sulfate) (ester), disodium salt (9CI), Phenol, 4,4'-(2-pyridylmethylene)di-, bis(hydrogen sulfate) (ester) disodium salt (8CI), 4,4'-(2-Pyridylmethylene)diphenol bis(hydrogen sulfate) disodium salt, DA 1773, Disodium bis[4-[(hydroxysulfonyl)oxy]phenyl](2-pyridyl)methane, Evacuol, Guttalax, Guttalax-Fher, LA 391, Laxoberal, Laxoberon, Neopax, Pico-Salax, Picolax, Picosulfate sodium, Picosulfol, Rapilax
InChI Formula InChI=1S/C18H15NO8S2.2Na/c20-28(21,22)26-15-8-4-13(5-9-15)18(17-3-1-2-12-19-17)14-6-10-16(11-7-14)27-29(23,24)25;;/h1-12,18H,(H,20,21,22)(H,23,24,25);;/q;2*+1/p-2
SMILES [Na+].[Na+].[O-]S(=O)(=O)Oc1ccc(cc1)C(c2ccc(OS(=O)(=O)[O-])cc2)c3ccccn3
IUPAC Name disodium;[4-[pyridin-2-yl-(4-sulfonatooxyphenyl)methyl]phenyl] sulfate
Molecular Weight (g/mol) 481.41
Quantity 250 mg
Formula Weight 480.9878 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.