missing translation for 'onlineSavingsMsg'
Learn More

Sodium tetrakis[3,5-bis(trifluoromethyl)phenyl]borate, 97%, may cont. 1-5% water

Product Code. 15495125
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15495125 25 g 25g
15475125 1 g 1g
15485125 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15495125 Supplier Thermo Scientific Alfa Aesar Supplier No. H30014.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sodium tetrakis[3,5-bis(trifluoromethyl)phenyl]borate, 97%, C32H12BF24Na, CAS Number-79060-88-1 | CAS: 79060-88-1 | C32H12BF24Na | 886.209 g/mol

Sodium tetrakis[3,5-bis(trifluoromethyl)phenyl]borate is a lipophilic additive of tetraphenylborate and it is used for solvent extraction of cations and solvent polymeric membrane electrodes and optodes. It is also used as an anionic phase-transfer catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium tetrakis[3,5-bis(trifluoromethyl)phenyl]borate is a lipophilic additive of tetraphenylborate and it is used for solvent extraction of cations and solvent polymeric membrane electrodes and optodes. It is also used as an anionic phase-transfer catalyst.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79060-88-1
Molecular Formula C32H12BF24Na
Molecular Weight (g/mol) 886.209
MDL Number MFCD00043323
InChI Key LTGMONZOZHXAHO-UHFFFAOYSA-N
Synonym sodium tetrakis 3,5-bis trifluoromethyl phenyl borate, nabarf, sodium tetrakis 3,5-bis trifluoromethyl phenyl boranuide, sodium2-sulfonatoethylmethacrylate, sodium tetrakis 3,5-bis trifluoromethyl phenyl bor, sodium tetrakis 3,5-bis-trifluoromethylphenyl borate, sodiotetrakis 3,5-bis trifluoromethyl phenyl boron v, sodium tetrakis 3,5-bis trifluoromethyl phenyl-borate, sodium tetrakis 3,5-bis trifluoromethyl phenyl borate 1-
PubChem CID 23681909
IUPAC Name sodium;tetrakis[3,5-bis(trifluoromethyl)phenyl]boranuide
SMILES [B-](C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F)(C2=CC(=CC(=C2)C(F)(F)F)C(F)(F)F)(C3=CC(=CC(=C3)C(F)(F)F)C(F)(F)F)C4=CC(=CC(=C4)C(F)(F)F)C(F)(F)F.[Na+]

Specifications

Melting Point 310°C (decomposition)
Quantity 25 g
Solubility Information Soluble in water.
Formula Weight 886.2
Percent Purity 97%
Assay may contain 1 to 5% water
Chemical Name or Material Sodium tetrakis[3,5-bis(trifluoromethyl)phenyl]borate
compliance-icons
Product Identifier
  • Sodium tetrakis¢3,5-bis(trifluoromethyl)phenyl!borate, may contain 1-5% water
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to in accordance with local/regional/national/international regulations..

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.