missing translation for 'onlineSavingsMsg'
Learn More

Sodium Thiophene-2-carboxylate, Mikromol™

Product Code. 30049148
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30049148 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30049148 Supplier Mikromol Supplier No. MM1081.00
 Restricted Item
Estimated Shipment: 31-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Sodium Thiophene-2-carboxylate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sodium Thiophene-2-carboxylate
CAS 25112-68-9
Analyte or Component Names Sodium Thiophene-2-carboxylate
Shipping Condition Room Temperature
Molecular Formula C5 H3 O2 S . Na
Certifications/Compliance ISO 17025
Synonym Sodium Thiophene-2-carboxylate, 2-Thiophenecarboxylic acid sodium salt, Sodium thiophene-2-carboxylate, Soufrane, Trophires
InChI Formula InChI=1S/C5H4O2S.Na/c6-5(7)4-2-1-3-8-4;/h1-3H,(H,6,7);/q;+1/p-1
SMILES [Na+].[O-]C(=O)c1cccs1
IUPAC Name sodium;thiophene-2-carboxylate
Molecular Weight (g/mol) 150.13
Quantity 250 mg
Formula Weight 149.9751 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.