missing translation for 'onlineSavingsMsg'
Learn More

Monkeypox Virus A29 Antibody (0036), Allophycocyanin, R&D Systems™

Product Code. 18613975 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18613975 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18613975 Supplier Novus Biologicals Supplier No. NBP318814APC
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Mouse Monoclonal Antibody

Monkeypox Virus A29 Monoclonal specifically detects Monkeypox Virus A29 in Virus samples. It is validated for ELISA∣Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Monkeypox Virus A29
Applications ELISA, Western Blot
Classification Monoclonal
Clone 0036
Conjugate APC
Dilution Western Blot, ELISA
Formulation PBS
Host Species Mouse
Immunogen This product is a recombinant monoclonal antibody made to recombinant Monkeypox Virus A29 Protein expressed from HEK293 cells.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4C in the dark.
Product Type Antibody
Form Purified
Isotype IgG1
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.