missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SOX2 Mouse, Clone: SOX2/1792, enQuireBio™
Mouse Monoclonal Antibody
Marca: EnquireBio 6657-MSM2-P1
Dettagli aggiuntivi : Weight : 0.01000kg
Specifica
SOX2 | |
Nuclear | |
SOX2/1792 | |
Unconjugated | |
NP_003097 | |
Mouse | |
34kDa | |
Protein A/G Purified | |
RUO | |
6657 | |
IgG2b κ |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
SOX2 | |
Recombinant human SOX2 protein fragment (aa176-305) | |
MCF-7 cells or Cervical carcinoma | |
100 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |