missing translation for 'onlineSavingsMsg'
Learn More

SOX3 Antibody (CL4725) [DyLight 755], Novus Biologicals™

Product Code. 30297294 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
Product Code. Quantity unitSize
30297294 0.1 mL 0.1mL
1 options
This item is not returnable. View return policy

Product Code. 30297294

Brand: Novus Biologicals NBP259060IR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

SOX3 Monoclonal antibody specifically detects SOX3 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen SOX3
Applications Western Blot
Classification Monoclonal
Clone CL4725
Conjugate DyLight 755
Formulation 50mM Sodium Borate
Gene Alias GHDX, MRGH, panhypopituitarism, PHP, PHPX, SOXB, SRY (sex determining region Y)-box 3, transcription factor SOX-3
Host Species Mouse
Immunogen This antibody was developed using a recombinant protein derived from P41225, with the exact immunogen sequence remaining proprietary.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6658
Target Species Human
Content And Storage Store at 4⪚C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.