missing translation for 'onlineSavingsMsg'
Learn More

SPAG4 Antibody [PE], Novus Biologicals™

Product Code. 30572711 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30572711 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30572711 Supplier Novus Biologicals Supplier No. NBP345857PE

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SPAG4 Polyclonal antibody specifically detects SPAG4 in Human,Mouse samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPAG4
Applications ELISA, Immunohistochemistry, Western Blot, Immunocytochemistry/Immunofluorescence
Classification Polyclonal
Conjugate PE
Formulation PBS
Gene Alias acrosomal protein ACR55, CT127, Outer dense fiber-associated protein SPAG4, Sad1 and UNC84 domain containing 4, sperm associated antigen 4, sperm tail protein, sperm-associated antigen 4 protein, SUN domain-containing protein 4, SUN4cancer/testis antigen 127
Host Species Rabbit
Immunogen sperm associated antigen 4. (Uniprot# Q9NPE6)
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6676
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.