missing translation for 'onlineSavingsMsg'
Learn More

SPANX-D Antibody [FITC], Novus Biologicals™

Product Code. 30580841 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30580841 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30580841 Supplier Novus Biologicals Supplier No. NBP345848F

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SPANX-D Polyclonal antibody specifically detects SPANX-D in Human samples. It is validated for ELISA,Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPANX-D
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate FITC
Formulation PBS
Gene Alias CT11.3;CT11.4;dJ171K16.1;SPANXC;SPANX-C;SPANXD;SPANX-D;SPANXE;SPANX-E;Sperm protein associated with the nucleus on the X chromosome D
Host Species Rabbit
Immunogen SPANX family, member E. (Uniprot# Q9BXN6)
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64648
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.