missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
SPARC-like 1/SPARCL1 Monoclonal specifically detects SPARC-like 1/SPARCL1 in Human samples. It is validated for ELISA, Sandwich ELISA.
Specifications
Specifications
| Antigen | SPARC-like 1/SPARCL1 |
| Applications | ELISA |
| Classification | Monoclonal |
| Conjugate | HRP |
| Dilution | Sandwich ELISA |
| Gene Alias | hevin, High endothelial venule protein, MAST 9, MAST9, PIG33, proliferation-inducing protein 33, SC1, SPARC-like 1 (hevin), SPARC-like 1 (mast9, hevin), SPARC-like protein 1 |
| Gene Symbols | SPARCL1 |
| Host Species | Mouse |
| Quantity | 15 Plates |
| Gene ID (Entrez) | 8404 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?