missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Spermatogensis associated protein 1 Antibody, mFluor Violet 610 SE, R&D Systems™
Shop All Bio Techne ProductsDescription
Spermatogensis associated protein 1 Polyclonal specifically detects Spermatogensis associated protein 1 in Human, Mouse samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot.
Specifications
Specifications
| Applications | ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Gene Alias | SP-2, SPAP1, SPATA1 spermatogenesis associated 1 |
| Host Species | Rabbit |
| Quantity | 0.1 mL |
| Regulatory Status | RUO |
| Research Discipline | Signal Transduction |
| Target Species | Mouse |
| Product Type | Antibody |
| Product Line | Novus |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?