missing translation for 'onlineSavingsMsg'
Learn More

SPHK1 Rabbit Recombinant Antibody, Proteintech

Rabbit Recombinant Antibody

129.00 € - 399.00 €

Specifications

Antigen SPHK1
Clone 2K12
Concentration 500 μg/mL
Applications Western Blot, Immunocytochemistry/Immunofluorescence, ELISA
Classification Recombinant Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30234231
View Documents
Proteintech
82609-1-RR-100UL
100 μL
ONLINE EXCLUSIVE PRICE
439.00 €
399.00 € / 100µL
Save 40.00 €
9% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30235486
View Documents
Proteintech
82609-1-RR-20UL
20 μL
ONLINE EXCLUSIVE PRICE
149.00 €
129.00 € / 20µL
Save 20.00 €
13% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The SPHK1 antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human SPHK1. This antibody recognizes human, mouse antigen. The SPHK1 antibody has been validated for the following applications: WB, IF/ICC, ELISA analysis.
Specifications

Specifications

SPHK1
500 μg/mL
Recombinant Monoclonal
Liquid
RUO
8877
Recombinant Protein
Primary
Store at -20°C. Stable for one year after shipment.
2K12
Western Blot, Immunocytochemistry/Immunofluorescence, ELISA
Unconjugated
Rabbit
Human, Mouse
SPHK
IgG
Protein A purification
Recombinant Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.