missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Spi-B Antibody (4G5), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-37458
This item is not returnable.
View return policy
Description
Spi-B Monoclonal specifically detects Spi-B in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| Spi-B | |
| Monoclonal | |
| Unconjugated | |
| Ascites with 0.03% Sodium Azide | |
| SPIB | |
| Purified recombinant fragment of human SPIB (AA: 200-252) expressed in E. Coli. | |
| Unpurified | |
| RUO | |
| 6689 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 4G5 | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000 | |
| SPI-B, Spi-B transcription factor (Spi-1/PU.1 related), transcription factor Spi-B | |
| Mouse | |
| 28.8 kDa | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| Ascites |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction