missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Spi-B Antibody (4G5), Novus Biologicals™
Mouse Monoclonal Antibody
196.00 € - 552.00 €
Specifications
| Antigen | Spi-B |
|---|---|
| Clone | 4G5 |
| Dilution | Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000 |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18647005
|
Novus Biologicals
NBP2-37458-0.025ml |
0.025 mL |
196.00 €
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18193599
|
Novus Biologicals
NBP2-37458 |
0.1 mL |
552.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Spi-B Monoclonal specifically detects Spi-B in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| Spi-B | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000 | |
| Monoclonal | |
| Ascites | |
| RUO | |
| Ascites with 0.03% Sodium Azide | |
| 6689 | |
| Purified recombinant fragment of human SPIB (AA: 200-252) expressed in E. Coli. | |
| Primary | |
| 28.8 kDa |
| 4G5 | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Human, Mouse | |
| SPI-B, Spi-B transcription factor (Spi-1/PU.1 related), transcription factor Spi-B | |
| SPIB | |
| IgG1 | |
| Unpurified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title