missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Spi-B Antibody (4G5), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-37458-0.025ml
This item is not returnable.
View return policy
Description
Spi-B Monoclonal specifically detects Spi-B in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| Spi-B | |
| Monoclonal | |
| Unconjugated | |
| Ascites with 0.03% Sodium Azide | |
| SPIB | |
| Purified recombinant fragment of human SPIB (AA: 200-252) expressed in E. Coli. | |
| Unpurified | |
| RUO | |
| 6689 | |
| Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 4G5 | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000 | |
| SPI-B, Spi-B transcription factor (Spi-1/PU.1 related), transcription factor Spi-B | |
| Mouse | |
| 28.8 kDa | |
| 0.025 mL | |
| Primary | |
| Human, Mouse | |
| Ascites |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction