missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Splicing Factor 1 Antibody [Alexa Fluor« 488], Novus Biologicals Biologicals™
Shop All Bio Techne ProductsDescription
Splicing Factor 1 Polyclonal antibody specifically detects Splicing Factor 1 in Human,Mouse samples. It is validated for ELISA,Western Blot
Specifications
Specifications
| Antigen | Splicing Factor 1 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 488 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | Mammalian branch point-binding protein, mBBP, splicing factor 1, Transcription factor ZFM1, ZFM1BBP, Zinc finger gene in MEN1 locus, Zinc finger protein 162D11S636, ZNF162MBBP |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-280 of human Splicing Factor 1 (NP_973727.1).,, Sequence:, MATGANATPLDFPSKKRKRSRWNQDTMEQKTVIPGMPTVIPPGLTREQERAYIVQLQIEDLTRKLRTGDLGIPPNPEDRSPSPEPIYNSEGKRLNTREFRTRKKLEEERHNLITEMVALNPDFKPPADYKPPATRVSDKVMIPQDEYPEINFVGLLIGPRGNTLKNIEKECNAKIMIRGKGSVKEGKVGRKDGQMLPGEDEPLHALVTANTMENVKKAVEQIRNILKQGIETPEDQNDLRKMQLRELARLNGTLREDDNRILRPWQSSETRSITNTTVCT |
| Purification Method | Affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?