missing translation for 'onlineSavingsMsg'
Learn More

SPRR1B Antibody [HRP], Novus Biologicals™

Product Code. 30580814 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30580814 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30580814 Supplier Novus Biologicals Supplier No. NBP345829H

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SPRR1B Polyclonal antibody specifically detects SPRR1B in Human samples. It is validated for ELISA,Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPRR1B
Applications ELISA, Immunohistochemistry
Classification Polyclonal
Conjugate HRP
Formulation PBS
Gene Alias 14.9 kDa pancornulin, CORNIFIN, cornifin B, cornifin-B, GADD33, MGC61901, small proline-rich protein 1B, Small proline-rich protein IB, SPR-IB, SPRR1
Host Species Rabbit
Immunogen small proline-rich protein 1B(cornifin). (Uniprot# P22528)
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6699
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.