missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SREBP1 Antibody (2A4) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Marque: Novus Biologicals NB600-582
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
SREBP1 Monoclonal specifically detects SREBP1 in Human, Mouse, Rat, Canine, Chicken, Hamster, Monkey, Golden Syrian Hamster samples. It is validated for Western Blot, Simple Western, Immunohistochemistry-Frozen, Knockout Validated.
Spécification
| SREBP1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1-2μg/mL, Simple Western 1:12.5, Immunohistochemistry-Frozen, Knockout Validated | |
| BHLHD1, Class D basic helix-loop-helix protein 1, SREBP 1c, SREBP-1, SREBP1bHLHd1SREBP-1c, sterol regulatory element binding protein-1, sterol regulatory element binding transcription factor 1, sterol regulatory element-binding protein 1, Sterol regulatory element-binding transcription factor 1 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, mouse, rat and Golden Syrian Hamster. Canine reactivity reported in scientific literature (PMID: 23720350). Hamster reactivity reported in scientific literature (PMID: 24393244). | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 κ |
| Western Blot, Immunohistochemistry (Frozen), Immunoassay | |
| 2A4 | |
| Unconjugated | |
| P36956 | |
| SREBF1 | |
| 6 His-tag fusion protein of human SREBP1 corresponding to amino acids 301-407. [UniProt# P36956] | |
| 0.1 mL | |
| Cancer, Cholesterol Metabolism, Chromatin Research, Diabetes Research, Lipid and Metabolism, Transcription Factors and Regulators | |
| 6720 | |
| Human, Mouse, Rat, Canine, Chicken, Hamster, Monkey, Golden Syrian Hamster | |
| Purified |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
For Research Use Only
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu