missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SREBP1 Antibody (2A4) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
199.00 € - 609.00 €
Specifications
| Antigen | SREBP1 |
|---|---|
| Clone | 2A4 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1-2μg/mL, Simple Western 1:12.5, Immunohistochemistry-Frozen, Knockout Validated |
| Applications | Western Blot, Immunohistochemistry (Frozen), Immunoassay |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18735063
|
Novus Biologicals
NB600-582SS |
0.025 mL |
199.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18725703
|
Novus Biologicals
NB600-582 |
0.1 mL |
609.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SREBP1 Monoclonal specifically detects SREBP1 in Human, Mouse, Rat, Canine, Chicken, Hamster, Monkey, Golden Syrian Hamster samples. It is validated for Western Blot, Simple Western, Immunohistochemistry-Frozen, Knockout Validated.Specifications
| SREBP1 | |
| 1 mg/mL | |
| Western Blot, Immunohistochemistry (Frozen), Immunoassay | |
| Unconjugated | |
| Mouse | |
| Cancer, Cholesterol Metabolism, Chromatin Research, Diabetes Research, Lipid and Metabolism, Transcription Factors and Regulators | |
| P36956 | |
| 6720 | |
| 6 His-tag fusion protein of human SREBP1 corresponding to amino acids 301-407. [UniProt# P36956] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 2A4 | |
| Western Blot 1-2μg/mL, Simple Western 1:12.5, Immunohistochemistry-Frozen, Knockout Validated | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat, Canine, Chicken, Hamster, Monkey, Golden Syrian Hamster | |
| BHLHD1, Class D basic helix-loop-helix protein 1, SREBP 1c, SREBP-1, SREBP1bHLHd1SREBP-1c, sterol regulatory element binding protein-1, sterol regulatory element binding transcription factor 1, sterol regulatory element-binding protein 1, Sterol regulatory element-binding transcription factor 1 | |
| SREBF1 | |
| IgG1 κ | |
| Protein G purified | |
| Human, mouse, rat and Golden Syrian Hamster. Canine reactivity reported in scientific literature (PMID: 23720350). Hamster reactivity reported in scientific literature (PMID: 24393244). |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title