missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Siglec-1/CD169 Antibody (HSn 7D2) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 18 publications
Brand: Novus Biologicals NB600-534
This item is not returnable.
View return policy
Description
Siglec-1/CD169 Monoclonal specifically detects Siglec-1/CD169 in Human, Mouse, Porcine, Virus samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, In vitro assay, Radioimmunoassay.
Specifications
| Siglec-1/CD169 | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot reported in scientific literature, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500. Use reported in scientific literature (PMID 26106049), Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 28794041), Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, In vitro assay, Radioimmunoassay, In vivo assay reported in scientific literature (PMID 18414664) | |
| CD169, FLJ00051, sialic acid binding Ig-like lectin 1, sialoadhesin, sialoadhesin | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human. Not yet tested in other species. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| HSn 7D2 | |
| Unconjugated | |
| Q9BZZ2 | |
| SIGLEC1 | |
| Full-length purified human Sialoadhesin (d1-4)Fc protein. [Swiss-Prot# Q9BZZ2] | |
| 0.1 mL | |
| Adaptive Immunity, Immunology | |
| 6614 | |
| Human, Mouse, Pig, Virus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction