missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Siglec-1/CD169 Antibody (HSn 7D2) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 18 publications
197.00 € - 561.00 €
Specifications
| Antigen | Siglec-1/CD169 |
|---|---|
| Clone | HSn 7D2 |
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot reported in scientific literature, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500. Use reported in scientific literature (PMID 26106049), Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 28794041), Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, In vitro assay, Radioimmunoassay, In vivo assay reported in scientific literature (PMID 18414664) |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18056975
|
Novus Biologicals
NB600-534SS |
0.025 mL |
197.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18750044
|
Novus Biologicals
NB600-534 |
0.1 mL |
561.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Siglec-1/CD169 Monoclonal specifically detects Siglec-1/CD169 in Human, Mouse, Porcine, Virus samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, In vitro assay, Radioimmunoassay.Specifications
| Siglec-1/CD169 | |
| 1.0 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Adaptive Immunity, Immunology | |
| Q9BZZ2 | |
| 6614 | |
| Full-length purified human Sialoadhesin (d1-4)Fc protein. [Swiss-Prot# Q9BZZ2] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| HSn 7D2 | |
| Western Blot reported in scientific literature, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500. Use reported in scientific literature (PMID 26106049), Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 28794041), Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, In vitro assay, Radioimmunoassay, In vivo assay reported in scientific literature (PMID 18414664) | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Pig, Virus | |
| CD169, FLJ00051, sialic acid binding Ig-like lectin 1, sialoadhesin, sialoadhesin | |
| SIGLEC1 | |
| IgG1 | |
| Protein G purified | |
| Human. Not yet tested in other species. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title