Learn More
SREBP1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 29 publications
Brand:  Novus Biologicals NB100-2215
 View more versions of this product
Product Code. 18745923
  • 594.00 € / 0.10mL
Description
SREBP1 Polyclonal specifically detects SREBP1 in Human, Mouse, Rat, Porcine, Bovine, Hamster, Plant samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Microarray.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
BHLHD1, Class D basic helix-loop-helix protein 1, SREBP 1c, SREBP-1, SREBP1bHLHd1SREBP-1c, sterol regulatory element binding protein-1, sterol regulatory element binding transcription factor 1, sterol regulatory element-binding protein 1, Sterol regulatory element-binding transcription factor 1
Affinity Purified
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
65 kDa
Pig