missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SREBP1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 29 publications
250.00 € - 594.00 €
Specifications
| Regulatory Status | RUO |
|---|---|
| Target Species | Human, Pig |
| Isotype | IgG |
| Content And Storage | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18701394
|
Novus Biologicals
NB100-2215SS |
0.025 mL |
250.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18745923
|
Novus Biologicals
NB100-2215 |
594.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
SREBP1 Polyclonal specifically detects SREBP1 in Human, Mouse, Rat, Porcine, Bovine, Hamster, Plant samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Microarray.Specifications
| RUO | |
| Human, Pig | |
| IgG | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title