missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ST8SIA-VI Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
280.00 € - 572.00 €
Specifications
| Antigen | ST8SIA-VI |
|---|---|
| Dilution | Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18491671
|
Novus Biologicals
NBP1-87042-25ul |
25 μL |
280.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18264667
|
Novus Biologicals
NBP1-87042 |
0.1 mL |
572.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ST8SIA-VI Polyclonal specifically detects ST8SIA-VI in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| ST8SIA-VI | |
| Polyclonal | |
| Rabbit | |
| Human | |
| 8-sialyltransferase), alpha-2,8-sialyltransferase 8F, alpha-2,8-sialyltransferase 8F variant 2, SIA8F, sialytransferase St8Sia VI, SIAT8F, ST8 alpha-N-acetyl-neuraminide alpha-2,8-sialyltransferase 6, ST8SiaVI, ST8SIA-VI | |
| ST8SIA6 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 338596 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:RILVEESREATHGTPAALRTLRSPATAVPRATNSTYLNEKSLQLTEKCKNLQYGIESFSNKTKGYSENDYLQIITDIQSCPWKRQAEEYANFRAKLASCCDAVQNFVVSQNNTPVGTNMSYEVESKKEIPIKKNIFHMFPVSQP | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title