missing translation for 'onlineSavingsMsg'
Learn More

STAM2 Antibody [mFluor Violet 450 SE], Novus Biologicals Biologicals™

Product Code. 30511079 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30511079 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30511079 Supplier Novus Biologicals Supplier No. NBP338372MFV450

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

STAM2 Polyclonal antibody specifically detects STAM2 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen STAM2
Applications ELISA, Western Blot, Immunocytochemistry/Immunofluorescence
Classification Polyclonal
Conjugate mFluor Violet 450 SE
Formulation 50mM Sodium Borate
Gene Alias DKFZp564C047, HBP, Hrs-binding protein, HSE1 homolog, signal transducing adapter molecule 2, signal transducing adaptor molecule (SH3 domain and ITAM motif) 2, STAM-2, STAM2A, STAM2B, STAM-like protein containing SH3 and ITAM domains 2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 376-525 of human STAM2 (NP_005834.4).,, Sequence:, KLHPPAHYPPASSGVPMQTYPVQSHGGNYMGQSIHQVTVAQSYSLGPDQIGPLRSLPPNVNSSVTAQPAQTSYLSTGQDTVSNPTYMNQNSNLQSATGTTAYTQQMGMSVDMSSYQNTTSNLPQLAGFPVTVPAHPVAQQHTNYHQQPLL
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10254
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.