missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STARD3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-21243-100ul
This item is not returnable.
View return policy
Description
STARD3 Polyclonal antibody specifically detects STARD3 in Human samples. It is validated for Western Blot
Specifications
| STARD3 | |
| Polyclonal | |
| Western Blot 0.04-0.4 ug/ml | |
| CAB1, es64, Metastatic lymph node gene 64 protein, metastatic lymph node protein 64, MLN 64, MLN64FLJ41370, Protein CAB1, StARD3, StAR-related lipid transfer (START) domain containing 3, stAR-related lipid transfer protein 3, START domain containing 3, START domain-containing protein 3, steroidogenic acute regulatory protein related | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: LPQEAEEERWYLAAQVAVARGPLLFSGALSEGQFYSPPESFAGSDNESDEEVAGKKSFSAQEREYIRQGKEATAVVDQILAQEENWKFEKNNEYGDTVYTIEVPFHGKTFILKTFLPCPAELVYQEVILQPERMVLWNK | |
| 100 μg | |
| Breast Cancer, Cancer, Lipid and Metabolism | |
| 10948 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS, pH 7.2, 40% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction