missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STARD3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
450.00 € - 648.00 €
Specifications
| Antigen | STARD3 |
|---|---|
| Dilution | Western Blot 0.04-0.4 ug/ml |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18622695
|
Novus Biologicals
NBP3-21243-100ul |
100 μg |
648.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18639744
|
Novus Biologicals
NBP3-21243-25ul |
25 μg |
450.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
STARD3 Polyclonal antibody specifically detects STARD3 in Human samples. It is validated for Western BlotSpecifications
| STARD3 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Breast Cancer, Cancer, Lipid and Metabolism | |
| PBS, pH 7.2, 40% glycerol | |
| 10948 | |
| IgG | |
| Affinity purified |
| Western Blot 0.04-0.4 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| CAB1, es64, Metastatic lymph node gene 64 protein, metastatic lymph node protein 64, MLN 64, MLN64FLJ41370, Protein CAB1, StARD3, StAR-related lipid transfer (START) domain containing 3, stAR-related lipid transfer protein 3, START domain containing 3, START domain-containing protein 3, steroidogenic acute regulatory protein related | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: LPQEAEEERWYLAAQVAVARGPLLFSGALSEGQFYSPPESFAGSDNESDEEVAGKKSFSAQEREYIRQGKEATAVVDQILAQEENWKFEKNNEYGDTVYTIEVPFHGKTFILKTFLPCPAELVYQEVILQPERMVLWNK | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title