missing translation for 'onlineSavingsMsg'
Learn More

Stargazin Antibody (S245-36), HRP, Novus Biologicals™

Product Code. 18156772 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18156772 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18156772 Supplier Novus Biologicals Supplier No. NBP222411H

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Stargazin Monoclonal specifically detects Stargazin in Human, Mouse, Rat samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Stargazin
Applications Western Blot
Classification Monoclonal
Clone S245-36
Conjugate HRP
Dilution Western Blot
Formulation PBS with No Preservative
Gene Alias calcium channel, voltage-dependent, gamma subunit 2, FLJ41437, MGC138502, MGC138504, Neuronal voltage-gated calcium channel gamma-2 subunit, stargazin, voltage-dependent calcium channel gamma-2 subunit
Gene Symbols CACNG2
Host Species Mouse
Immunogen Fusion protein amino acids 203-323 (Cytoplasmic C-terminus) of rat TARPGamma2
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 10369
Test Specificity Detects approx 35-55kDa. Does not cross-react with TARPGamma3.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.