missing translation for 'onlineSavingsMsg'
Learn More

START domain containing 7 Antibody [Janelia Fluor™ 669], Novus Biologicals™

Product Code. 30568701 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30568701 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30568701 Supplier Novus Biologicals Supplier No. NBP345800JF669

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

START domain containing 7 Polyclonal antibody specifically detects START domain containing 7 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen START domain containing 7
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Janelia Fluor 669
Formulation 50mM Sodium Borate
Gene Alias Gestational trophoblastic tumor protein 1, GTT1stAR-related lipid transfer protein 7, mitochondrial, StARD7, StAR-related lipid transfer (START) domain containing 7, START domain containing 7, START domain-containing protein 7
Host Species Rabbit
Immunogen StAR-related lipid transfer(START) domain containing 7. (Uniprot# Q9NQZ5)
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56910
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.