missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Staufen Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
369.00 € - 593.00 €
Specifications
| Antigen | Staufen |
|---|---|
| Dilution | Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:200 - 1:500 |
| Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18166318
|
Novus Biologicals
NBP2-38615 |
0.1 mL |
593.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18650646
|
Novus Biologicals
NBP2-38615-25ul |
25 μL |
369.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Staufen Polyclonal specifically detects Staufen in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| Staufen | |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| double-stranded RNA-binding protein Staufen homolog 1, FLJ25010, RNA binding protein (Drosophila), RNA-binding protein), staufen, RNA binding protein, homolog 1 (Drosophila) | |
| STAU1 | |
| IgG | |
| Affinity Purified |
| Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| O95793 | |
| 6780 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: TRPSEQLDYLSRVQGFQVEYKDFPKNNKNEFVSLINCSSQPPLISHGIGKDVESCHDMAALNILKLLSELDQQSTEMPRTGNGPMSVCG | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title